APO455832850 Apollo Scientific Ltd

3-(Fur-2-yl)benzaldehyde

  • MDL number MFCD03990461
  • Cas Number : 85553-52-2
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-furyl)benzaldehyde
Inchi Code 1S/C11H8O2/c12-8-9-3-1-4-10(7-9)11-5-2-6-13-11/h1-8H
Inchi Key BTAANNDAXIYWAN-UHFFFAOYSA-N

Description

Synonym 2-(3-Formylphenyl)furan
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fur-2-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fur-2-ylbenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart