APO455832856 Apollo Scientific Ltd

Methyl 5-methoxy-2-nitrobenzoate

  • MDL number MFCD00662902
  • Cas Number : 2327-45-9
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-57
Synonym 4-Methoxy-2-(methoxycarbonyl)nitrobenzene, 3-(Methoxycarbonyl)-4-nitroanisole
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-methoxy-2-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-methoxy-2-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart