APO455832859 Apollo Scientific Ltd

6-Bromo-1-methyl-1H-benzimidazole

  • MDL number MFCD09027271
  • Cas Number : 53484-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-203
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-bromo-1-methyl-1h-benzimidazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-1-methyl-1h-benzimidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart