APO455832863 Apollo Scientific Ltd

4-(Methylsulphonyl)benzonitrile

  • MDL number MFCD00216489
  • Cas Number : 22821-76-7
  • Molecular Weight : 181.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 137-142
Iupac Name 4-(methylsulfonyl)benzonitrile
Inchi Code 1S/C8H7NO2S/c1-12(10,11)8-4-2-7(6-9)3-5-8/h2-5H,1H3
Inchi Key FARXIDYHJAANGP-UHFFFAOYSA-N

Description

Synonym 4-Cyanophenyl methyl sulphone
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 137-142
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart