APO455832865 Apollo Scientific Ltd

alpha-Methyl-4-(methylsulphonyl)benzylamine

  • MDL number MFCD01319250
  • Cas Number : 387350-90-5
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 65-67
Iupac Name 1-[4-(methylsulfonyl)phenyl]ethanamine
Inchi Code 1S/C9H13NO2S/c1-7(10)8-3-5-9(6-4-8)13(2,11)12/h3-7H,10H2,1-2H3
Inchi Key XJAMLZUKCSRXAF-UHFFFAOYSA-N

Description

Synonym 1-[4-(Methylsulphonyl)phenyl]ethylamine, 4-(1-Aminoethyl)phenyl methyl sulphone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 65-67
Product Link https://store.apolloscientific.co.uk/product/alpha-methyl-4-methylsulphonylbenzylamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/alpha-methyl-4-methylsulphonylbenzylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart