APO455832866 Apollo Scientific Ltd

2-Amino-3'-methoxybiphenyl hydrochloride

  • MDL number MFCD06739419
  • Cas Number : 1082749-15-2
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3-Methoxyphenyl)aniline hydrochloride, 3'-Methoxy-[1,1'-biphenyl]-2-amine hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-methoxybiphenyl-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-methoxybiphenyl-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart