APO455832867 Apollo Scientific Ltd

2'-Methoxy-[1,1'-biphenyl]-3-amine hydrochloride

  • MDL number MFCD06739423
  • Cas Number : 96923-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(2-Methoxyphenyl)aniline hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-11-biphenyl-3-amine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-11-biphenyl-3-amine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart