APO455832868 Apollo Scientific Ltd

3-Amino-4'-methoxybiphenyl hydrochloride

  • MDL number MFCD09787507
  • Cas Number : 53059-28-2
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4'-methoxy[1,1'-biphenyl]-3-amine hydrochloride
Inchi Code 1S/C13H13NO.ClH/c1-15-13-7-5-10(6-8-13)11-3-2-4-12(14)9-11;/h2-9H,14H2,1H3;1H
Inchi Key DLIGUYMDOVTEIJ-UHFFFAOYSA-N

Description

Synonym 4'-Methoxy-[1,1'-biphenyl]-3-amine hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-methoxybiphenyl-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-methoxybiphenyl-hydrochloride
pictograms GHS07
hstatements H315;H319;H335;H302
Availability details for
Quantity
  • Availability
  • Add to Cart