APO455832869 Apollo Scientific Ltd

3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride

  • MDL number MFCD07772428
  • Cas Number : 207287-79-4
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(3-Methoxyphenyl)aniline hydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methoxy-11-biphenyl-4-amine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxy-11-biphenyl-4-amine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart