APO455832870 Apollo Scientific Ltd

2'-Methoxy-[1,1'-biphenyl]-4-carboxaldehyde

  • MDL number MFCD01631913
  • Cas Number : 421553-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Synonym 4-(2-Methoxyphenyl)benzaldehyde, 2-(4-Formylphenyl)anisole, 4-Formyl-2'-methoxybiphenyl
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-11-biphenyl-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-11-biphenyl-4-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart