APO455832871 Apollo Scientific Ltd

3'-Methoxy-[1,1'-biphenyl]-3-carboxaldehyde

  • MDL number MFCD03424626
  • Cas Number : 126485-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(3-Methoxyphenyl)benzaldehyde, 3-(3-Formylphenyl)anisole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27194

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27194
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart