APO455832872 Apollo Scientific Ltd

3'-Methoxy-[1,1'-biphenyl]-4-carboxaldehyde

  • MDL number MFCD03424619
  • Cas Number : 209863-09-2
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(3-Methoxyphenyl)benzaldehyde, 4'-Formyl-3-methoxybiphenyl, 3-(4-Formylphenyl)anisole
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methoxy-11-biphenyl-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P280;P260
MSDS https://store.apolloscientific.co.uk/product/3-methoxy-11-biphenyl-4-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart