APO455832875 Apollo Scientific Ltd

1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one

  • MDL number MFCD04039102
  • Cas Number : 192863-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27209

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27209
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart