APO455832878 Apollo Scientific Ltd

5-(3-Methoxyphenyl)-2-furoic acid

  • MDL number MFCD05022489
  • Cas Number : 54022-96-7
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-170
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-3-methoxyphenyl-2-furoic-acid

Safety Information

signalword Danger
pcodes P260;P280;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/5-3-methoxyphenyl-2-furoic-acid
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart