APO455832879 Apollo Scientific Ltd

Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride

  • MDL number MFCD07366482
  • Cas Number : 343985-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3'-Amino-3-(methoxycarbonyl)biphenyl hydrochloride, 3-[3-(Methoxycarbonyl)phenyl]aniline hydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27219

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27219
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart