APO455832881 Apollo Scientific Ltd

2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride

  • MDL number MFCD06739417
  • Cas Number : 1203-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27223

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27223
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart