APO455832884 Apollo Scientific Ltd

4-Amino-2'-methylbiphenyl hydrochloride

  • MDL number MFCD06739442
  • Cas Number : 811842-48-5
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2'-methyl[1,1'-biphenyl]-4-amine hydrochloride
Inchi Code 1S/C13H13N.ClH/c1-10-4-2-3-5-13(10)11-6-8-12(14)9-7-11;/h2-9H,14H2,1H3;1H
Inchi Key OBZHYWKQOJFWEL-UHFFFAOYSA-N

Description

Synonym 2'-Methyl-[1,1'-biphenyl]-4-amine hydrochloride, 4-(2-Methylphenyl)aniline hydrochloride
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-methylbiphenyl-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-methylbiphenyl-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart