APO455832888 Apollo Scientific Ltd

2'-Methyl-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD03424589
  • Cas Number : 168618-44-8
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2'-methyl[1,1'-biphenyl]-3-carboxylic acid
Inchi Code 1S/C14H12O2/c1-10-5-2-3-8-13(10)11-6-4-7-12(9-11)14(15)16/h2-9H,1H3,(H,15,16)
Inchi Key IAZJDWZUCUHVPQ-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-2'-methylbiphenyl, 3-(2-Methylphenyl)benzoic acid, 2-(3-Carboxyphenyl)toluene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-11-biphenyl-3-carboxylic-acid_OR7514

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-11-biphenyl-3-carboxylic-acid_OR7514
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart