APO455832889 Apollo Scientific Ltd

2'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD03424591
  • Cas Number : 5748-43-6
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2'-methyl[1,1'-biphenyl]-4-carboxylic acid
Inchi Code 1S/C14H12O2/c1-10-4-2-3-5-13(10)11-6-8-12(9-7-11)14(15)16/h2-9H,1H3,(H,15,16)
Inchi Key NDNIPPKLIDCYGD-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-2'-methylbiphenyl, 4-(2-Methylphenyl)benzoic acid, 2-(4-Carboxyphenyl)toluene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-11-biphenyl-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-11-biphenyl-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart