APO455832890 Apollo Scientific Ltd

3'-Methyl-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD00558892
  • Cas Number : 158619-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-3'-methylbiphenyl, 3-(3-Methylphenyl)benzoic acid, 3-(3-Carboxyphenyl)toluene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-methyl-11-biphenyl-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-11-biphenyl-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart