APO455832891 Apollo Scientific Ltd

3'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD03424590
  • Cas Number : 5728-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(3-Methylphenyl)benzoic acid, 3-(4-Carboxyphenyl)toluene, 4-Carboxy-3'-methylbiphenyl
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27245

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27245
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart