APO455832893 Apollo Scientific Ltd

2-(5-Methylfur-2-yl)aniline hydrochloride

  • MDL number MFCD01191726
  • Cas Number : 329187-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2-Aminophenyl)-5-methylfuran hydrochloride
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27257

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27257
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart