APO455832897 Apollo Scientific Ltd

4-(5-Methylfur-2-yl)benzaldehyde

  • MDL number MFCD04039125
  • Cas Number : 400748-10-9
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-Formylphenyl)-5-methylfuran
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-5-methylfur-2-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-5-methylfur-2-ylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart