APO455832901 Apollo Scientific Ltd

3,5-Dichloropyridin-4(1H)-one

  • MDL number MFCD00051673
  • Cas Number : 17228-70-5
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 325
Iupac Name 3,5-dichloro-4(1H)-pyridinone
Inchi Code 1S/C5H3Cl2NO/c6-3-1-8-2-4(7)5(3)9/h1-2H,(H,8,9)
Inchi Key YKCQWIYRLMNGMO-UHFFFAOYSA-N

Description

Synonym 3,5-Dichloro-1,4-dihydro-4-oxopyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 325
Product Link https://store.apolloscientific.co.uk/product/35-dichloropyridin-41h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloropyridin-41h-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart