APO455832909 Apollo Scientific Ltd

3,5-Dichloro-4-hydroxybenzenesulphonyl chloride

  • MDL number MFCD00171551
  • Cas Number : 13432-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Chlorosulphonyl)-2,6-dichlorophenol
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14180

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14180
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart