APO455832919 Apollo Scientific Ltd

2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propanoic acid

  • MDL number MFCD04039159
  • Cas Number : 5162-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(1,2-Dihydro-2-oxoquinolin-4-yl)-DL-alanine, 2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propionic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27308

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27308
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart