APO455832920 Apollo Scientific Ltd

1-Chloro-6,6-dimethylhept-2-en-4-yne

  • MDL number MFCD04039160
  • Cas Number : 126764-17-8
  • Molecular Weight : 156.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-1-chloro-6,6-dimethyl-2-hepten-4-yne
Inchi Code 1S/C9H13Cl/c1-9(2,3)7-5-4-6-8-10/h4,6H,8H2,1-3H3/b6-4+
Inchi Key ZIXABMZBMHDFEZ-GQCTYLIASA-N

Description

Synonym 6,6-Dimethylhept-2-en-4-yn-1-yl chloride
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-chloro-66-dimethylhept-2-en-4-yne

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-chloro-66-dimethylhept-2-en-4-yne
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart