APO455832921 Apollo Scientific Ltd

2-[4-(5-Bromopyrimidin-2-yl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD03452782
  • Cas Number : 849021-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-99
Synonym 1-(5-Bromopyrimidin-2-yl)-4-(2-hydroxyethyl)piperazine, 5-Bromo-2-[4-(2-hydroxyethyl)piperazin-1-yl]pyrimidine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-4-5-bromopyrimidin-2-ylpiperazin-1-ylethan-1-ol

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-4-5-bromopyrimidin-2-ylpiperazin-1-ylethan-1-ol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart