APO455832922 Apollo Scientific Ltd

4-Hydroxybenzamidine hydrochloride

  • MDL number MFCD00136405
  • Cas Number : 38148-63-9
  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-226
Synonym 4-Hydroxybenzenecarboximidamide hydrochloride, 4-Carbamimidoylphenol hydrochloride
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxybenzamidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxybenzamidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart