APO455832930 Apollo Scientific Ltd

2-(4-Chlorophenoxy)malonaldehyde

  • MDL number MFCD03426063
  • Cas Number : 849021-40-5
  • Molecular Weight : 198.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 116-118
Iupac Name 2-(4-chlorophenoxy)malonaldehyde
Inchi Code 1S/C9H7ClO3/c10-7-1-3-8(4-2-7)13-9(5-11)6-12/h1-6,9H
Inchi Key PXSIXELVDKBAKQ-UHFFFAOYSA-N

Description

Synonym (4-Chlorophenoxy)propane-1,3-dial
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 116-118
Product Link https://store.apolloscientific.co.uk/product/2-4-chlorophenoxymalonaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-chlorophenoxymalonaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart