APO455832935 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-nitrobenzaldehyde

  • MDL number MFCD04037929
  • Cas Number : 849035-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-5-(methylsulphonyl)nitrobenzene, 4-Formyl-3-nitrophenyl methyl sulphone
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27347

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27347
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart