APO455832938 Apollo Scientific Ltd

2'-Chloroacetanilide

  • MDL number MFCD00045169
  • Cas Number : 533-17-5
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 87-89
Iupac Name N-(2-chlorophenyl)acetamide
Inchi Code 1S/C8H8ClNO/c1-6(11)10-8-5-3-2-4-7(8)9/h2-5H,1H3,(H,10,11)
Inchi Key KNVQTRVKSOEHPU-UHFFFAOYSA-N

Description

Synonym N-(2-Chlorophenyl)acetamide
Purity 98%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 87-89
Product Link https://store.apolloscientific.co.uk/product/2-chloroacetanilide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloroacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart