APO455832940 Apollo Scientific Ltd

Pyrimidine-5-carboxaldehyde

  • MDL number MFCD03426065
  • Cas Number : 10070-92-5
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 39-43
Iupac Name 5-pyrimidinecarbaldehyde
density 1.23
Inchi Code 1S/C5H4N2O/c8-3-5-1-6-4-7-2-5/h1-4H
Inchi Key FREJAOSUHFGDBW-UHFFFAOYSA-N

Description

Synonym 5-Formylpyrimidine, 5-Formyl-1,3-diazine
Density 1.23
Flash Point 80-82/4mm
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 39-43
Product Link https://store.apolloscientific.co.uk/product/pyrimidine-5-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyrimidine-5-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart