APO455832943 Apollo Scientific Ltd

1-Benzylhomopiperazin-5-one

  • MDL number MFCD00225259
  • Cas Number : 55186-89-5
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 115-117
Iupac Name 1-benzyl-1,4-diazepan-5-one
Inchi Code 1S/C12H16N2O/c15-12-6-8-14(9-7-13-12)10-11-4-2-1-3-5-11/h1-5H,6-10H2,(H,13,15)
Inchi Key DKNOPZCYIDBMNY-UHFFFAOYSA-N

Description

Synonym 1-Benzyl-1,4-diazepan-5-one, 1-Benzyl-5-oxo-1,4-diazepane
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 115-117
Product Link https://store.apolloscientific.co.uk/product/1-benzylhomopiperazin-5-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-benzylhomopiperazin-5-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart