APO455832945 Apollo Scientific Ltd

5-(3-Chlorophenyl)-1,3-oxazole

  • MDL number MFCD00084974
  • Cas Number : 89808-76-4
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 46-48
Iupac Name 5-(3-chlorophenyl)-1,3-oxazole
Inchi Code 1S/C9H6ClNO/c10-8-3-1-2-7(4-8)9-5-11-6-12-9/h1-6H
Inchi Key NJVHPUNGYNOCQN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 46-48
Product Link https://store.apolloscientific.co.uk/product/5-3-chlorophenyl-13-oxazole

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS https://store.apolloscientific.co.uk/product/5-3-chlorophenyl-13-oxazole
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart