APO455832959 Apollo Scientific Ltd

Methyl 4-ethylbenzoate

  • MDL number MFCD00221477
  • Cas Number : 7364-20-7
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 105-107/15mm
Product Link https://store.apolloscientific.co.uk/product/methyl-4-ethylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-ethylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart