APO455832961 Apollo Scientific Ltd

4-(Cyclopropylamino)piperidine, N1-BOC protected

  • MDL number MFCD03426074
  • Cas Number : 179557-01-8
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-(cyclopropylamino)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(cyclopropylamino)piperidine
Purity 95
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyclopropylaminopiperidine-n1-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyclopropylaminopiperidine-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart