APO455832964 Apollo Scientific Ltd

2-(tert-Butyl)-N-(piperidin-4-yl)-1H-indol-5-amine

  • MDL number MFCD01319470
  • Cas Number : 397842-07-8
  • Molecular Weight : 271.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Synonym 2-(tert-Butyl)-5-[(piperidin-4-yl)amino]-1H-indole, 4-{[2-(tert-Butyl)-1H-indol-5-yl]amino}piperidine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-tert-butyl-n-piperidin-4-yl-1h-indol-5-amine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-tert-butyl-n-piperidin-4-yl-1h-indol-5-amine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart