APO455832967 Apollo Scientific Ltd

4-Bromo-3-[(dimethylamino)methyl]phenol

  • MDL number MFCD06657798
  • Cas Number : 848848-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-N,N-dimethyl-5-hydroxybenzylamine, 1-(2-Bromo-5-hydroxyphenyl)-N,N-dimethylmethylamine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36513

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36513
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart