APO455832990 Apollo Scientific Ltd

4-Hydrazinobenzenesulphonamide hydrochloride

  • MDL number MFCD00052262
  • Cas Number : 17852-52-7
  • Molecular Weight : 223.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 217-219(dec.)
Iupac Name 4-hydrazinobenzenesulfonamide hydrochloride
Inchi Code 1S/C6H9N3O2S.ClH/c7-9-5-1-3-6(4-2-5)12(8,10)11;/h1-4,9H,7H2,(H2,8,10,11);1H
Inchi Key IKEURONJLPUALY-UHFFFAOYSA-N

Description

Synonym 4-Sulphamoylphenylhydrazine hydrochloride, 4-(Aminosulphonyl)phenylhydrazine hydrochloride
Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 217-219(dec.)
Product Link https://store.apolloscientific.co.uk/product/4-hydrazinobenzenesulphonamide-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydrazinobenzenesulphonamide-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart