APO455832996 Apollo Scientific Ltd

N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride

  • MDL number MFCD04039172
  • Cas Number : 849020-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-191
Synonym 3-[Acetyl(methyl)amino]benzylamine hydrochloride, 3-(Aminomethyl)-N-methylacetanilide hydrochloride
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27405

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27405
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart