APO455833000 Apollo Scientific Ltd

2-Isopropoxybenzylamine

  • MDL number MFCD04037890
  • Cas Number : 227199-51-1
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-isopropoxyphenyl)methanamine
Inchi Code 1S/C10H15NO/c1-8(2)12-10-6-4-3-5-9(10)7-11/h3-6,8H,7,11H2,1-2H3
Inchi Key RJWAKPHKRZMFHG-UHFFFAOYSA-N

Description

Synonym (2-Isopropoxyphenyl)methylamine, {[2-(Prop-2-yl)oxy]phenyl}methylamine
Purity 95%
Flash Point 53-56/0.07mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-isopropoxybenzylamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-isopropoxybenzylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart