APO455833002 Apollo Scientific Ltd

4-Isopropoxybenzylamine

  • MDL number MFCD04037892
  • Cas Number : 21244-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(Prop-2-yl)oxy]benzylamine, (4-Isopropoxyphenyl)methylamine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 74/0.22mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27410

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27410
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart