APO455833004 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-nitroaniline

  • MDL number MFCD00718367
  • Cas Number : 21731-56-6
  • Molecular Weight : 216.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 194-196
Iupac Name 4-(methylsulfonyl)-2-nitroaniline
Inchi Code 1S/C7H8N2O4S/c1-14(12,13)5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3
Inchi Key NDZFWKZHVAUUTN-UHFFFAOYSA-N

Description

Synonym 2-Amino-5-(methylsulphonyl)nitrobenzene, 4-Amino-3-nitrophenyl methyl sulphone
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 194-196
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart