APO455833006 Apollo Scientific Ltd

4-{3-[4-(Methylsulphonyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine

  • MDL number MFCD03426082
  • Cas Number : 849925-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-131
Synonym 3-[4-(Methylsulphonyl)phenyl]-5-(piperidin-4-yl)-1,2,4-oxadiazole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27420

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27420
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart