APO455833008 Apollo Scientific Ltd

5-Bromo-2-methoxy-4-(methylthio)pyrimidine

  • MDL number MFCD07781219
  • Cas Number : 59549-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36570

Safety Information

pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36570
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart