APO455833012 Apollo Scientific Ltd

3-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyridine

  • MDL number MFCD03426078
  • Cas Number : 849925-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-83
Synonym 5-(Piperidin-4-yl)-3-(pyridin-3-yl)-1,2,4-oxadiazole, 4-[3-(Pyridin-3-yl)-1,2,4-oxadiazol-5-yl]piperidine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27426

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27426
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart