APO455833016 Apollo Scientific Ltd

4-[3-(Thien-2-yl)-1,2,4-oxadiazol-5-yl]piperidine

  • MDL number MFCD03426076
  • Cas Number : 244272-35-3
  • Molecular Weight : 235.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-71
Synonym 5-(Piperidin-4-yl)-3-(thien-2-yl)-1,2,4-oxadiazole, 2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]thiophene
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-thien-2-yl-124-oxadiazol-5-ylpiperidine

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-3-thien-2-yl-124-oxadiazol-5-ylpiperidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart