APO455833017 Apollo Scientific Ltd

3-[Acetyl(methyl)amino]propanoic acid

  • MDL number MFCD04037898
  • Cas Number : 58706-66-4
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 91-92
Iupac Name N-acetyl-N-methyl-beta-alanine
Inchi Code 1S/C6H11NO3/c1-5(8)7(2)4-3-6(9)10/h3-4H2,1-2H3,(H,9,10)
Inchi Key CUAUMKXJKVIJHG-UHFFFAOYSA-N

Description

Synonym 3-[Acetyl(methyl)amino]propionic acid, N-Acetyl-N-methyl-beta-alanine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91-92
Product Link https://store.apolloscientific.co.uk/product/3-acetylmethylaminopropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-acetylmethylaminopropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart