APO455833020 Apollo Scientific Ltd

4-Bromo-2-(methylsulphonyl)benzoic acid

  • MDL number MFCD04037926
  • Cas Number : 142994-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-224
Synonym 5-Bromo-2-carboxyphenyl methyl sulphone
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27438

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27438
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart